O-Benzylhydroxylamine hydrochloride ≥98.0%
Supplier: TCI America
B1079-250G
B1079-25G
B1079-5G
TCB1079-250GEA
930.62
CAD
TCB1079-250G
TCB1079-025G
TCB1079-5G
O-Benzylhydroxylamine hydrochloride ≥98.0%
O-Benzylhydroxylamine hydrochloride
CAS Number: 2687-43-6
MDL Number: MFCD00012952
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₇H₁₀ClNO MW: 159.62 g/mol |
MDL Number:
MFCD00012952 CAS Number: 2687-43-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...