Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

B1045-1G B1045-100MG
TCB1045-001GEA 1015.94 CAD
TCB1045-001G TCB1045-100MG
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98.0% (by titrimetric analysis)
Chloro(1,5-cyclooctadiene)rhodium (I) dimer

CAS Number: 12092-47-6
MDL Number: MFCD00012415
Molecular Formula: C16H24Cl2Rh2
Molecular Weight: 493.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Bis(1,5-cyclooctadiene)dirhodium(I) Dichloride, 1,5-Cyclooctadiene Rhodium(I) Chloride Dimer, [Rh(COD)Cl]2

Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
MDL Number: MFCD00012415
CAS Number: 12092-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f3f382320f965496a4bda888a0c0b36515b03a1&vid=5f3f382320f965496a4bda888a0c0b36515b03a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR