1-(tert-Butyldimethylsilyl)imidazole ≥98.0%

Supplier: TCI America

Synonyms: 1-(tert-Butyldimethylsilyl)-1H-imidazole

B1043-5G B1043-1G
TCB1043-005GEA 234.79 CAD
TCB1043-005G TCB1043-001G
1-(tert-Butyldimethylsilyl)imidazole ≥98.0%
1-(tert-Butyldimethylsilyl)imidazole

[tert-Butyldimethylsilylating Agent]
CAS Number: 54925-64-3
MDL Number: MFCD00011682
Molecular Formula: C9H18N2Si
Molecular Weight: 182.34
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 81
Specific Gravity (20/20): 0.86

Formula: C₉H₁₈N₂Si
MW: 182.34 g/mol
Boiling Pt: 155 °C
Density: 0.86 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00011682
CAS Number: 54925-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d81ee92bc62728e9e00aebaecc53edb94ee85eb&vid=2d81ee92bc62728e9e00aebaecc53edb94ee85eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR