1,4-Bis(2-methylstyryl)benzene ≥99.0%

Supplier: TCI America

B1024-5G B1024-25G
TCB1024-005GEA 79 CAD
TCB1024-005G 76219-330
1,4-Bis(2-methylstyryl)benzene ≥99.0%
1,4-Bis(2-methylstyryl)benzene

[Solute for Liquid Scintillation Counting]
CAS Number: 13280-61-0
MDL Number: MFCD00008529
Molecular Formula: C24H22
Molecular Weight: 310.44
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 179


Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: 1,4-Di(2-methylstyryl)benzene

Formula: C₂₄H₂₂
Melting Pt: 179 °C
MDL Number: MFCD00008529
CAS Number: 13280-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8044becd6b1aef158e9b288eead7d708e5c9752e&vid=8044becd6b1aef158e9b288eead7d708e5c9752e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR