Bathocuproin sulfonate disodium salt
Supplier: TCI America
B0985-100MG
B0985-1G
TCB0985-100MGEA
77.42
CAD
TCB0985-100MG
TCB0985-001G
Bathocuproin sulfonate disodium salt
Bathocuproin sulphonate disodium salt
[for Determination of Cu in Blood]
CAS Number: 52698-84-7
MDL Number: MFCD00011679
Molecular Formula: C26H20N2O6S2
Molecular Weight: 564.54
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthrolinedisulfonic Acid Disodium Salt, Bathocuproinedisulfonic Acid Disodium Salt
Formula:
C₂₆H₁₈N₂Na₂O₆S₂ |
MDL Number:
MFCD00011679 CAS Number: 52698-84-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...