4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98.0%
Supplier: TCI America
Synonyms:
4-Benzoyl-5-methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one
B0976-5G
B0976-25G
TCB0976-005GEA
172.22
CAD
TCB0976-005G
TCB0976-025G
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98.0%
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone
CAS Number: 4551-69-3
MDL Number: MFCD00003136
Molecular Formula: C17H14N2O2
Molecular Weight: 278.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118
Formula:
C₁₇H₁₄N₂O₂ Melting Pt: 118 °C |
MDL Number:
MFCD00003136 CAS Number: 4551-69-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...