Bis(4-chlorobutyl)ether ≥98.0%

Supplier: TCI America

B0929-25ML B0929-500ML
TCB0929-025MLEA 52.93 CAD
TCB0929-025ML TCB0929-500ML
Bis(4-chlorobutyl)ether ≥98.0%
Bis(4-chlorobutyl)ether

CAS Number: 6334-96-9
MDL Number: MFCD00019006
Molecular Formula: C8H16Cl2O
Molecular Weight: 199.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 4-Chlorobutyl Ether, 4,4'-Dichlorobutyl Ether, 4,4'-Dichlorodibutyl Ether

Formula: C₈H₁₆Cl₂O
Boiling Pt: 132 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00019006
CAS Number: 6334-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8a3644695f782ebacdd75dd6db86e539b72cdc5&vid=b8a3644695f782ebacdd75dd6db86e539b72cdc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR