n-Butyl phenyl ether ≥99.0%
Supplier: TCI America
B0927-500ML
B0927-25ML
TCB0927-500MLEA
1350.9
CAD
TCB0927-500ML
TCB0927-025ML
n-Butyl phenyl ether ≥99.0%
n-Butyl phenyl ether
CAS Number: 1126-79-0
MDL Number: MFCD00009438
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Melting point (°C): -19
Flash Point (°C): 82
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: Butoxybenzene
Formula:
C₁₀H₁₄O Boiling Pt: 210 °C Melting Pt: –19 °C Density: 0.94 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00009438 CAS Number: 1126-79-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...