5-Bromo-3-nitrosalicylaldehyde ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromo-2-hydroxy-3-nitrobenzaldehyde

B0908-1G B0908-5G
TCB0908-1GEA 58.46 CAD
TCB0908-1G TCB0908-5G
5-Bromo-3-nitrosalicylaldehyde ≥97.0% (by GC, titration analysis)
5-Bromo-3-nitrosalicylaldehyde

CAS Number: 16634-88-1
MDL Number: MFCD00130110
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 5-Bromo-2-hydroxy-3-nitrobenzaldehyde

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 125 °C
MDL Number: MFCD00130110
CAS Number: 16634-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3bab5a92c0bf251d3c153858ce918008b7ba679&vid=c3bab5a92c0bf251d3c153858ce918008b7ba679&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR