O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-Ser(Bzl)-OH
B0861-1G
B0861-5G
TCB0861-001GEA
50.56
CAD
TCB0861-001G
TCB0861-005G
O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)
O-Benzyl-L-serine
CAS Number: 4726-96-9
MDL Number: MFCD00065937
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 7.3 deg (C=2, 1mol/L HCl)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Ser(Bzl)-OH
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol |
MDL Number:
MFCD00065937 CAS Number: 4726-96-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...