4,4'-Dihydroxydiphenylmethane ≥99.0%
Supplier: TCI America
B0819-5G
B0819-25G
B0819-100G
TCB0819-005GEA
126.4
CAD
TCB0819-005G
TCB0819-025G
TCB0819-100G
4,4'-Dihydroxydiphenylmethane ≥99.0%
4,4'-Dihydroxydiphenylmethane
CAS Number: 620-92-8
MDL Number: MFCD00002385
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 160
Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Bis(p-hydroxyphenyl)methane, 4,4'-Methylenediphenol
Formula:
C₁₃H₁₂O₂ Melting Pt: 160 °C |
MDL Number:
MFCD00002385 CAS Number: 620-92-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...