Ethyl-2-butylacetoacetate ≥96.0%
Supplier: TCI America
Synonyms:
2-Acetylhexanoic Acid Ethyl Ester, 2-Butylacetoacetic Acid Ethyl Ester, Ethyl 2-acetylhexanoate, Ethyl 2-Acetylhexanoate
CAS Number: 1540-29-0
MDL Number: MFCD00026888
Molecular Formula: C10H18O3
Molecular Weight: 186.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 224
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 2-Acetylhexanoic Acid Ethyl Ester, Ethyl 2-Acetylhexanoate, 2-Butylacetoacetic Acid Ethyl Ester
Formula:
C₁₀H₁₈O₃ MW: 186.25 g/mol Boiling Pt: 224 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00026888 CAS Number: 1540-29-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...