Sodium 2-Bromoethanesulfonate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
B0589-25G
B0589-500G
TCB0589-025GEA
99.54
CAD
TCB0589-025G
TCB0589-500G
Sodium 2-Bromoethanesulfonate ≥97.0% (by titrimetric analysis)
Sodium 2-Bromoethanesulfonate
CAS Number: 4263-52-9
MDL Number: MFCD00007530
Molecular Formula: C2H5BrO3S
Molecular Weight: 211.00
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 286
Other Product Names / Synonyms: 2-Bromoethanesulfonic Acid Sodium Salt
Formula:
C₂H₅BrO₃S Melting Pt: 286 °C |
MDL Number:
MFCD00007530 CAS Number: 4263-52-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...