Biurea
Supplier: TCI America
B0512-25G
B0512-5G
TCB0512-025GEA
883.22
CAD
TCB0512-025G
TCB0512-5G
Biurea
Biurea
CAS Number: 110-21-4
MDL Number: MFCD00025398
Molecular Formula: C2H6N4O2
Molecular Weight: 118.10
Form: Crystal
Color: White
Formula:
C₂H₆N₄O₂ |
MDL Number:
MFCD00025398 CAS Number: 110-21-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...