2,2'-Biquinoline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B0470-5G B0470-1G
TCB0470-005GEA 263.51 CAD
TCB0470-005G TCB0470-001G
2,2'-Biquinoline ≥98.0% (by HPLC, titration analysis)
2,2'-Biquinoline

CAS Number: 119-91-5
MDL Number: MFCD00006740
Molecular Formula: C18H12N2
Molecular Weight: 256.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 192


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: Cuproine, 2,2'-Diquinolyl

Formula: C₁₈H₁₂N₂
MW: 256.31 g/mol
Melting Pt: 192 °C
MDL Number: MFCD00006740
CAS Number: 119-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c45088a4473fc65d616129dc19de2283e65045de&vid=c45088a4473fc65d616129dc19de2283e65045de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR