Benzyl phenyl ether ≥98.0%
Supplier: TCI America
B0433-25G
B0433-500G
TCB0433-025GEA
83.74
CAD
TCB0433-025G
TCB0433-500G
Benzyl phenyl ether ≥98.0%
Benzyl phenyl ether
CAS Number: 946-80-5
MDL Number: MFCD00020660
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 39
Flash Point (°C): 113
Storage Temperature: 0-10°C
Formula:
C₁₃H₁₂O Boiling Pt: 288 °C Melting Pt: 39 °C Flash Pt: 113 °C |
MDL Number:
MFCD00020660 CAS Number: 946-80-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...