Diethyl benzylidenemalonate ≥96.0%
Supplier: TCI America
Synonyms:
Diethyl Benzalmalonate, Benzalmalonic Acid Diethyl Ester, Benzylidenemalonic Acid Diethyl Ester
B0420-25G
B0420-5G
TCB0420-025GEA
66.36
CAD
TCB0420-025G
TCB0420-5G
Diethyl benzylidenemalonate ≥96.0%
Diethyl benzylidenemalonate
CAS Number: 5292-53-5
MDL Number: MFCD00009149
Molecular Formula: C14H16O4
Molecular Weight: 248.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: Benzylidenemalonic Acid Diethyl Ester, Diethyl Benzalmalonate, Benzalmalonic Acid Diethyl Ester
Formula:
C₁₄H₁₆O₄ MW: 248.28 g/mol Density: 1.10 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00009149 CAS Number: 5292-53-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...