Leuco malachite green ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B0419-1G
B0419-25G
TCB0419-001GEA
48.24
CAD
TCB0419-001G
TCB0419-025G
Leuco malachite green ≥98.0% (by titrimetric analysis)
Leuco malachite green
CAS Number: 129-73-7
MDL Number: MFCD00008315
Molecular Formula: C23H26N2
Molecular Weight: 330.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 102
Other Product Names / Synonyms: 4,4'-Benzylidenebis(N,N-dimethylaniline), 4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-Tetramethyldiaminotriphenylmethane
Formula:
C₂₃H₂₆N₂ Melting Pt: 102 °C |
MDL Number:
MFCD00008315 CAS Number: 129-73-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...