2-Naphthyl phenyl ketone ≥98.0%

Supplier: TCI America

Synonyms: 2-Benzoylnaphthalene

B0301-25G B0301-5G
TCB0301-25GEA 772.62 CAD
TCB0301-25G TCB0301-005G
2-Naphthyl phenyl ketone ≥98.0%
2-Naphthyl phenyl ketone

CAS Number: 644-13-3
MDL Number: MFCD00004106
Molecular Formula: C17H12O
Molecular Weight: 232.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 82


Other Product Names / Synonyms: 2-Benzoylnaphthalene

Formula: C₁₇H₁₂O
MW: 232.28 g/mol
Boiling Pt: 225 °C
Melting Pt: 82 °C
MDL Number: MFCD00004106
CAS Number: 644-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9695a0ddfd1bd6e69e7665c1e5fe60913ca8c486&vid=9695a0ddfd1bd6e69e7665c1e5fe60913ca8c486&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR