L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-Ser-OMe.HCl
B0267-25G
B0267-5G
TCB0267-025GEA
79
CAD
TCB0267-025G
TCB0267-5G
L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-serine methyl ester hydrochloride
CAS Number: 5680-80-8
MDL Number: MFCD00063680
Molecular Formula: C4H9NO3
Molecular Weight: 155.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 161
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Ser-OMe.HCl
Formula:
C₄H₉NO₃ MW: 155.58 g/mol Melting Pt: 161 °C |
MDL Number:
MFCD00063680 CAS Number: 5680-80-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...