N(α)-Cbz-L-histidine ≥99.0%

Supplier: TCI America

Synonyms: Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH

B0265-5G B0265-25G B0265-1G
TCB0265-5GEA 213.3 CAD
TCB0265-5G TCB0265-25G TCB0265-001G
N(α)-Cbz-L-histidine ≥99.0%
N(α)-Cbz-L-histidine

CAS Number: 14997-58-1
MDL Number: MFCD00065960
Molecular Formula: C14H15N3O4
Molecular Weight: 289.29
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: -23.5 deg (C=6, 6mol/L HCl)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-L-histidine, Z-His-OH

Formula: C₁₄H₁₅N₃O₄
MW: 289.3 g/mol
Melting Pt: 163 °C
MDL Number: MFCD00065960
CAS Number: 14997-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6da8b50853346f093bb994861fed8f3d51142af&vid=d6da8b50853346f093bb994861fed8f3d51142af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR