Benzyl butyl ether ≥95.0%
Supplier: TCI America
B0239-25G
B0239-5G
TCB0239-25GEA
238.58
CAD
TCB0239-25G
TCB0239-5G
Benzyl butyl ether ≥95.0%
Benzyl butyl ether
CAS Number: 588-67-0
MDL Number: MFCD00039954
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 91
Specific Gravity (20/20): 0.92
Formula:
C₁₁H₁₆O Boiling Pt: 220 °C Density: 0.92 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00039954 CAS Number: 588-67-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...