Benzhydrylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B0124-25ML B0124-500ML B0124-100ML
TCB0124-025MLEA 60.04 CAD
TCB0124-025ML TCB0124-500ML TCB0124-100ML
Benzhydrylamine ≥97.0% (by GC, titration analysis)
Benzhydrylamine

CAS Number: 91-00-9
MDL Number: MFCD00008059
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 304
Melting point (°C): 12
Flash Point (°C): 110
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: alpha-Aminodiphenylmethane, 1,1-Diphenylmethylamine

Formula: C₁₃H₁₃N
Boiling Pt: 304 °C
Melting Pt: 12 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008059
CAS Number: 91-00-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7398a243c0a30c6a3e14fe7e7a498f4a7bf1196&vid=f7398a243c0a30c6a3e14fe7e7a498f4a7bf1196&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR