Ethyl phenylglyoxylate ≥96.0%

Supplier: TCI America

B0109-25ML B0109-5ML
TCB0109-025MLEA 121.66 CAD
TCB0109-025ML TCB0109-5ML
Ethyl phenylglyoxylate ≥96.0%
Ethyl phenylglyoxylate

CAS Number: 1603-79-8
MDL Number: MFCD00009120
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Benzoylformic Acid Ethyl Ester, Ethyl Phenylglyoxylate, Phenylglyoxylic Acid Ethyl Ester

Formula: C₁₀H₁₀O₃
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00009120
CAS Number: 1603-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea03bfacc6c4fd8c2fde2e99e3d9769e6edcaaf7&vid=ea03bfacc6c4fd8c2fde2e99e3d9769e6edcaaf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR