Benzo[h]quinoline ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B0088-25G B0088-5G
TCB0088-25GEA 374.46 CAD
TCB0088-25G TCB0088-5G
Benzo[h]quinoline ≥99.0% (by GC, titration analysis)
Benzo[h]quinoline

CAS Number: 230-27-3
MDL Number: MFCD00004984
Molecular Formula: C13H9N
Molecular Weight: 179.22
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 338
Melting point (°C): 50


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 4-Azaphenanthrene, 7,8-Benzoquinoline, alpha-Naphthoquinoline

Formula: C₁₃H₉N
Boiling Pt: 338 °C
Melting Pt: 50 °C
MDL Number: MFCD00004984
CAS Number: 230-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22f40142cb0ec1f07e08893d4646db82ba24abb6&vid=22f40142cb0ec1f07e08893d4646db82ba24abb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR