Benzohydroxamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0061-5G B0061-25G
TCB0061-5GEA 71.86 CAD
TCB0061-5G TCB0061-025G
Benzohydroxamic acid ≥98.0% (by titrimetric analysis)
Benzohydroxamic acid

CAS Number: 495-18-1
MDL Number: MFCD00002109
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: N-Hydroxybenzamide

Formula: C₇H₇NO₂
Melting Pt: 124 °C
MDL Number: MFCD00002109
CAS Number: 495-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bd7b9cbc13c13ab54a5ed8e6470d031b5ea87b7&vid=9bd7b9cbc13c13ab54a5ed8e6470d031b5ea87b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR