Sodium benzenesulfonate ≥96.0% (by titrimetric analysis)
Supplier: TCI America
B0035-25G
B0035-500G
B0035-100G
TCB0035-025GEA
63.2
CAD
TCB0035-025G
TCB0035-500G
TCB0035-100G
Sodium benzenesulfonate ≥96.0% (by titrimetric analysis)
Sodium benzenesulfonate
CAS Number: 515-42-4
MDL Number: MFCD00065179
Molecular Formula: C6H6O3S
Molecular Weight: 180.15
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Other Product Names / Synonyms: Benzenesulfonic Acid Sodium Salt
Formula:
C₆H₆O₃S |
MDL Number:
MFCD00065179 CAS Number: 515-42-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...