Benzal diacetate ≥95.0%

Supplier: TCI America

Synonyms: Benzylidene Diacetate

B0010-25G B0010-500G
TCB0010-025GEA 131.8 CAD
TCB0010-025G TCB0010-500G
Benzal diacetate ≥95.0%
Benzal diacetate

CAS Number: 581-55-5
MDL Number: MFCD00059206
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 45


Other Product Names / Synonyms: Benzylidene Diacetate

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 154 °C
Melting Pt: 45 °C
MDL Number: MFCD00059206
CAS Number: 581-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66e63836fe12a7bce58962767270fc7708158f96&vid=66e63836fe12a7bce58962767270fc7708158f96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR