ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0%

Supplier: TCI America

Synonyms: ABD-F, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulfonamide, 4-Fluoro-7-sulfobenzofurazan, ammonium salt, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide, 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide

A5597-100MG A5597-500MG
TCA5597-100MGEA 543.52 CAD
TCA5597-100MG TCA5597-500MG
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0%
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]

[HPLC Labeling Reagent for Determination of Thiols]
CAS Number: 91366-65-3
MDL Number: MFCD00036687
Molecular Formula: C6H4FN3O3S
Molecular Weight: 217.17
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 146


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole

Formula: C₆H₄FN₃O₃S
MW: 217.17 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00036687
CAS Number: 91366-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03f49dabdf9bfe5abb0c7a6f92d52f31032cefa4&vid=03f49dabdf9bfe5abb0c7a6f92d52f31032cefa4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR