O-(4-Nitrobenzyl)hydroxylamine hydrochloride ≥99.0%

Supplier: TCI America

Synonyms: O-(4-Nitrobenzyl)-hydroxylammonium chloride

A5532-1G A5532-5G
TCA5532-001GEA 142.2 CAD
TCA5532-001G TCA5532-5G
O-(4-Nitrobenzyl)hydroxylamine hydrochloride ≥99.0%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride

[for HPLC Labeling]
CAS Number: 2086-26-2
MDL Number: MFCD00012954
Molecular Formula: C7H8N2O3
Molecular Weight: 204.61
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 210

Formula: C₇H₉ClN₂O₃
MW: 204.61 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00012954
CAS Number: 2086-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5b736caa18d16c485db0e49f5f40094a15dab9&vid=670d3dab143bf732c4286826f7ed8c35d3680e74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR