Fluoral-P (=4-amino-3-penten-2-one) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-3-penten-2-one, 4-Aminopent-3-en-2-one

A5350-1G A5350-5G
TCA5350-001GEA 69.52 CAD
TCA5350-001G TCA5350-005G
Fluoral-P (=4-amino-3-penten-2-one) ≥98.0% (by GC, titration analysis)
Fluoral-P (=4-amino-3-penten-2-one)

[Fluorimetric reagent for aldehydes.]
CAS Number: 1118-66-7
MDL Number: MFCD00043715
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 214
Melting point (°C): 41
Storage Temperature: <0°C


Other Product Names / Synonyms: 4-Amino-3-penten-2-one

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 214 °C
Melting Pt: 41 °C
MDL Number: MFCD00043715
CAS Number: 1118-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38e5dadde37a49744491246ab5dc2ac2ca595e77&vid=38e5dadde37a49744491246ab5dc2ac2ca595e77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR