1-(2-Thiazolylazo)-2-naphthol ≥99.0%

Supplier: TCI America

Synonyms: NSC-139021, TAN, 1-(2-Thiazoylazo)-2-naphthol

A5003-1G A5003-5G
TCA5003-001GEA 118.97 CAD
TCA5003-001G TCA5003-005G
1-(2-Thiazolylazo)-2-naphthol ≥99.0%
1-(2-Thiazolylazo)-2-naphthol

[Spectrophotometric reagent for transition metals]
CAS Number: 1147-56-4
MDL Number: MFCD00021611
Molecular Formula: C13H9N3OS
Molecular Weight: 255.30
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 141


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 1-(2-Thiazolylazo)-2-naphthol

Formula: C₁₃H₉N₃OS
MW: 255.30 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00021611
CAS Number: 1147-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb81f8c13c5293411a9db1b344305d63e0182c14&vid=0abda3bad63d4039610d295e6b27db7ea14051ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR