4-(2-Thiazolylazo)resorcinol ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A5002-5G A5002-1G
TCA5002-005GEA 323.9 CAD
TCA5002-005G TCA5002-001G
4-(2-Thiazolylazo)resorcinol ≥99.0% (by titrimetric analysis)
4-(2-Thiazolylazo)resorcinol

[Metal indicator and spectrophotometric reagent for transition metals]
CAS Number: 2246-46-0
MDL Number: MFCD00005322
Molecular Formula: C9H7N3O2S
Molecular Weight: 221.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 209


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 4-(2-Thiazolylazo)resorcinol

Formula: C₉H₇N₃O₂S
Melting Pt: 209 °C
MDL Number: MFCD00005322
CAS Number: 2246-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49c6fae784a1d49e189cd494d643df7e8e5c4dd3&vid=e2cd0682cd571cd973c82cbede4ed9c0102e8874&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR