5-Acetylindan ≥98.0%
Supplier: TCI America
Synonyms:
1-(2,3-Dihydro-1H-inden-5-yl)ethan-1-one, 1-(2,3-Dihydro-1H-inden-5-yl)ethanone
A2289-25G
A2289-5G
TCA2289-25GEA
597.24
CAD
TCA2289-25G
TCA2289-5G
5-Acetylindan ≥98.0%
5-Acetylindan
CAS Number: 4228-10-8
MDL Number: MFCD00021246
Molecular Formula: C11H12O
Molecular Weight: 160.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Specific Gravity (20/20): 1.06
Lambda max.: 258 nm (EtOH)
Formula:
C₁₁H₁₂O MW: 160.22 g/mol Boiling Pt: 85 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00021246 CAS Number: 4228-10-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...