4-Amino-3-nitropyridine ≥98.0%
Supplier: TCI America
Synonyms:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
A2260-25G
A2260-5G
TCA2260-25GEA
499.12
CAD
TCA2260-25G
TCA2260-5G
4-Amino-3-nitropyridine ≥98.0%
4-Amino-3-nitropyridine
CAS Number: 1681-37-4
MDL Number: MFCD00160304
Molecular Formula: C5H5N3O2
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 205
Formula:
C₅H₅N₃O₂ Melting Pt: 205 °C |
MDL Number:
MFCD00160304 CAS Number: 1681-37-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...