3-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2259-25G
A2259-5G
TCA2259-25GEA
243
CAD
TCA2259-25G
TCA2259-5G
3-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
3-Aminobenzyl alcohol
CAS Number: 1877-77-6
MDL Number: MFCD00007817
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 94
Other Product Names / Synonyms: 3-(Hydroxymethyl)aniline
Formula:
C₇H₉NO Melting Pt: 94 °C |
MDL Number:
MFCD00007817 CAS Number: 1877-77-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...