3-Acetylcoumarin ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetyl-2H-chromen-2-one

A2200-5G A2200-25G
TCA2200-5GEA 72.68 CAD
TCA2200-5G TCA2200-25G
3-Acetylcoumarin ≥98.0%
3-Acetylcoumarin

CAS Number: 3949-36-8
MDL Number: MFCD00006853
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 123
Lambda max.: 302 nm (EtOH)

Formula: C₁₁H₈O₃
MW: 188.18
Melting Pt: 123 °C
MDL Number: MFCD00006853
CAS Number: 3949-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5521f47ce156cd84cdfbb60d39d30b3a30b2cd57&vid=5521f47ce156cd84cdfbb60d39d30b3a30b2cd57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR