6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Aminopyridine-2-carboxylic acid

A2188-1G A2188-5G
TCA2188-1GEA 75.84 CAD
TCA2188-1G TCA2188-5G
6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)
6-Aminopicolinic acid

CAS Number: 23628-31-1
MDL Number: MFCD00233711
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 315


Other Product Names / Synonyms: 6-Aminopicolinic Acid

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 315 °C
MDL Number: MFCD00233711
CAS Number: 23628-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3757533f75846ac5437fe52888ea482c9299bb22&vid=3757533f75846ac5437fe52888ea482c9299bb22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR