(4-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2182-5G
A2182-25G
TCA2182-5GEA
72.68
CAD
TCA2182-5G
TCA2182-25G
(4-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(4-Acetylphenoxy)acetic acid
CAS Number: 1878-81-5
MDL Number: MFCD00014363
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174
Formula:
(C₆H₄(C(O)CH₃))OCH₂COOH Melting Pt: 174 °C |
MDL Number:
MFCD00014363 CAS Number: 1878-81-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...