4-Amino-3-fluorobenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Amino-3-fluorobenzoic Acid

A2165-5G A2165-1G
TCA2165-5GEA 75.84 CAD
TCA2165-5G TCA2165-1G
4-Amino-3-fluorobenzoic acid ≥98.0%
4-Amino-3-fluorobenzoic acid

CAS Number: 455-87-8
MDL Number: MFCD01660374
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₇H₆FNO₂
MDL Number: MFCD01660374
CAS Number: 455-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcecf57fae6ba98c978996d8f6e1b62f0c90faef&vid=679141a4c846a49ce794fc7e370a98a03e09c17f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR