4-Amino-2-fluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2163-5G A2163-1G
TCA2163-5GEA 378.57 CAD
TCA2163-5G TCA2163-1G
4-Amino-2-fluorophenol ≥98.0% (by GC, titration analysis)
4-Amino-2-fluorophenol

CAS Number: 399-96-2
MDL Number: MFCD00671760
Molecular Formula: C6H6FNO
Molecular Weight: 127.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 164


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-4-hydroxyaniline

Formula: C₆H₆FNO
Melting Pt: 164 °C
MDL Number: MFCD00671760
CAS Number: 399-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9fee6f982f5c1827464584de640337e1bf74155&vid=d9fee6f982f5c1827464584de640337e1bf74155&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR