2-Amino-5-bromo-3-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2154-1G A2154-5G
TCA2154-1GEA 44.56 CAD
TCA2154-1G TCA2154-5G
2-Amino-5-bromo-3-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromo-3-methylpyridine

CAS Number: 3430-21-5
MDL Number: MFCD00068232
Molecular Formula: C6H7BrN2
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 94


Other Product Names / Synonyms: 2-Amino-5-bromo-3-picoline

Formula: C₆H₇BrN₂
Melting Pt: 94 °C
MDL Number: MFCD00068232
CAS Number: 3430-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e25069d992bc845a1830e2cf2e5f528b39cc6798&vid=e25069d992bc845a1830e2cf2e5f528b39cc6798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR