D-α-Aminoadipic acid ≥95.0%

Supplier: TCI America

Synonyms: D-2-Aminoadipic acid, (R)-2-Aminohexanedioic acid, H-D-Aad-OH

A2153-1G A2153-5G
TCA2153-1GEA 82.16 CAD
TCA2153-1G TCA2153-5G
D-α-Aminoadipic acid ≥95.0%
D-α-Aminoadipic acid

CAS Number: 7620-28-2
MDL Number: MFCD00063118
Molecular Formula: C6H11NO4
Molecular Weight: 161.16
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: -24 deg (C=1, 6mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Aminohexanedioic Acid

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00063118
CAS Number: 7620-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9a3ce5024f648610bc5d62e04c0521055369d08&vid=f9a3ce5024f648610bc5d62e04c0521055369d08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR