4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98.0%

Supplier: TCI America

Synonyms: Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline

A2151-25G A2151-5G
TCA2151-25GEA 259.97 CAD
TCA2151-25G TCA2151-5G
4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98.0%
4-Amino-2-chloro-6,7-dimethoxyquinazoline

CAS Number: 23680-84-4
MDL Number: MFCD00051734
Molecular Formula: C10H10ClN3O2
Molecular Weight: 239.66
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 302


Other Product Names / Synonyms: 2-Chloro-6,7-dimethoxy-4-quinazolinamine

Formula: C₁₀H₁₀ClN₃O₂
Melting Pt: 302 °C
MDL Number: MFCD00051734
CAS Number: 23680-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03dfa0f969a09f087ac7fd2eaa194a33e6042e60&vid=03dfa0f969a09f087ac7fd2eaa194a33e6042e60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR