3-Acetamidotetrahydro-2-thiophenone ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: Citiolone, 2-Acetamido-4-mercaptobutyric Acid gamma-Thiolactone, N-Acetyl-DL-homocystein Thiolactone, N-(Tetrahydro-2-oxo-3-thienyl)acetamide, N-(2-Oxotetrahydrothiophen-3-yl)acetamide

A2144-25G A2144-5G
TCA2144-25GEA 361.68 CAD
TCA2144-25G TCA2144-5G
3-Acetamidotetrahydro-2-thiophenone ≥98.0% (by GC, total nitrogen)
3-Acetamidotetrahydro-2-thiophenone

CAS Number: 1195-16-0
MDL Number: MFCD00005480
Molecular Formula: C6H9NO2S
Molecular Weight: 159.20
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 110


Product Categories: Bioscience, Functionality Creating Reagents
Other Product Names / Synonyms: N-Acetyl-DL-homocystein Thiolactone, 2-Acetamido-4-mercaptobutyric Acid gamma-Thiolactone, N-(Tetrahydro-2-oxo-3-thienyl)acetamide, Citiolone

Formula: C₆H₉NO₂S
MW: 159.20 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00005480
CAS Number: 1195-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=415198ec8e9fb057785a10cbcaac88da7a0dbbf9&vid=415198ec8e9fb057785a10cbcaac88da7a0dbbf9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR