2-Amino-3-chloropyrazine ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine

A2143-1G A2143-5G
TCA2143-1GEA 39.5 CAD
TCA2143-1G TCA2143-5G
2-Amino-3-chloropyrazine ≥98.0%
2-Amino-3-chloropyrazine

CAS Number: 6863-73-6
MDL Number: MFCD04114305
Molecular Formula: C4H4ClN3
Molecular Weight: 129.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168

Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 168 °C
MDL Number: MFCD04114305
CAS Number: 6863-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63963ec1a076a133f85426a90bc7a4393ee5428c&vid=63963ec1a076a133f85426a90bc7a4393ee5428c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR