4-Amino-6-chloropyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2136-5G A2136-1G
TCA2136-5GEA 208.24 CAD
TCA2136-5G TCA2136-1G
4-Amino-6-chloropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-6-chloropyrimidine

CAS Number: 5305-59-9
MDL Number: MFCD00053576
Molecular Formula: C4H4ClN3
Molecular Weight: 129.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 220

Formula: C₄H₄ClN₃
Melting Pt: 220 °C
MDL Number: MFCD00053576
CAS Number: 5305-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32ba001967cd1c9bf7186a653dbdb4752c782f9&vid=e32ba001967cd1c9bf7186a653dbdb4752c782f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR