N-Phenyl-o-phenylenediamine ≥98.0%
Supplier: TCI America
Synonyms:
N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine
A2129-5G
A2129-25G
TCA2129-5GEA
55.3
CAD
TCA2129-5G
TCA2129-25G
N-Phenyl-o-phenylenediamine ≥98.0%
N-Phenyl-o-phenylenediamine
CAS Number: 534-85-0
MDL Number: MFCD00007685
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 80
Other Product Names / Synonyms: N-Phenyl-o-phenylenediamine
Formula:
C₁₂H₁₂N₂ Boiling Pt: 108 °C Melting Pt: 80 °C |
MDL Number:
MFCD00007685 CAS Number: 534-85-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...