4-Aminobenzamidine dihydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
A2115-25G
A2115-5G
TCA2115-25GEA
505.6
CAD
TCA2115-25G
TCA2115-5G
4-Aminobenzamidine dihydrochloride ≥98.0% (by total nitrogen basis)
4-Aminobenzamidine dihydrochloride
CAS Number: 2498-50-2
MDL Number: MFCD00013001
Molecular Formula: C7H9N3
Molecular Weight: 208.09
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 291
Storage Temperature: 0-10°C
Formula:
C₇H₁₁Cl₂N₃ Melting Pt: 291 °C |
MDL Number:
MFCD00013001 CAS Number: 2498-50-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...