3-Amino-5-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Amino-5-phenylpyrazole
A2102-1G
A2102-5G
TCA2102-1GEA
44.56
CAD
TCA2102-1G
TCA2102-5G
3-Amino-5-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-5-phenyl-1H-pyrazole
CAS Number: 1572-10-7
MDL Number: MFCD00191749
Molecular Formula: C9H9N3
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 126
Formula:
C₉H₉N₃ MW: 159.19 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD00191749 CAS Number: 1572-10-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...